Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIBRA Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB185275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185275 Supplier Novus Biologicals Supplier No. NBP282101V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIBRA Polyclonal antibody specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen KIBRA
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1
Host Species Rabbit
Immunogen KIBRA antibody was raised against an 18 amino acid peptide near the amino terminus of human KIBRA. The immunogen is located within amino acids 60 - 110 of KIBRA. Amino Acid Squence: KTTQIEDPRVQWRREQEH
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23286
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.