Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIBRA Antibody (OTI1H8), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271732A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271732A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271732A48 Supplier Novus Biologicals Supplier No. NBP271732AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIBRA Monoclonal specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen KIBRA
Applications Western Blot
Classification Monoclonal
Clone OTI1H8
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1
Gene Symbols WWC1
Host Species Mouse
Immunogen Full length recombinant protein of human KIBRA (NP_056053) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23286.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.