Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIBRA Antibody (OTI1H8), PE, Novus Biologicals™
SDP

Catalog No. NB288675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288675 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288675 Supplier Novus Biologicals Supplier No. NBP271732PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIBRA Monoclonal antibody specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen KIBRA
Applications Western Blot
Classification Monoclonal
Clone OTI1H8
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1
Host Species Mouse
Immunogen Full length human recombinant protein of human WWC1 (NP_056053) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23286
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.