Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIDINS220 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291258 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291258 Supplier Novus Biologicals Supplier No. NBP150034PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIDINS220 Polyclonal antibody specifically detects KIDINS220 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen KIDINS220
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias kinase D-interacting substrate, 220kDa
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1400 and 1450 of human Ankyrin Repeat-Rich Membrane Spanning using the numbering given in entry NP_065789.1 (GeneID 57498).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 57498
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.