Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF14 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292316 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292316 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292316 Supplier Novus Biologicals Supplier No. NB100254APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF14 Polyclonal specifically detects KIF14 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen KIF14
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias KIAA0042MGC142302, kinesin family member 14, kinesin-like protein KIF14
Host Species Rabbit
Immunogen Maps to a region between residues 900 and 950 of human Kinesin Family Member 14 using the numbering given in entry NP_055690.1 (GeneID 9928).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9928
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.