Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF14 Antibody, Novus Biologicals™
SDP

Catalog No. NB11040678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040678 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040678 Supplier Novus Biologicals Supplier No. NB11040678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF14 Polyclonal specifically detects KIF14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen KIF14
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunocytochemistry
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_055690.1
Gene Alias KIAA0042MGC142302, kinesin family member 14, kinesin-like protein KIF14
Gene Symbols KIF14
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1600 and the C-terminus (residue 1648) of human Kinesin Family Member 14 using the numbering given in entry NP_055690.1 (GeneID 9928).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9928
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.