Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF17 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001319 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001319 Supplier Novus Biologicals Supplier No. NB6001319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF17 Polyclonal specifically detects KIF17 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.

Specifications

Antigen KIF17
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Microarray
Formulation 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gene Alias KIAA1405KIF3XKIF17 variant protein, KIF17B, KIF3-related motor protein, kinesin family member 17, kinesin-like protein KIF17
Gene Symbols KIF17
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH, corresponding to amino acids 589-606 of Mouse KIF17.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 57576
Test Specificity KIF17
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.