Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | KIF19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15816820
![]() |
Novus Biologicals
NBP15816820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP158168
![]() |
Novus Biologicals
NBP158168 |
100 μL |
Each for $487.50
|
|
|||||
Description
KIF19 Polyclonal specifically detects KIF19 in Human samples. It is validated for Western Blot.Specifications
KIF19 | |
Polyclonal | |
Rabbit | |
Q8N1X8 | |
124602 | |
Synthetic peptides corresponding to FLJ37300 The peptide sequence was selected from the N terminal of FLJ37300. Peptide sequence EVSMSYLEIYNEMIRDLLNPSLGYLELREDSKGVIQVAGITEVSTINAKE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ37300, KIF19A, kinesin family member 19, kinesin-like protein KIF19 | |
KIF19 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title