Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF21A Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094276 20 μL
NB094275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094276 Supplier Novus Biologicals Supplier No. NBP30438920UL

Rabbit Polyclonal Antibody

KIF21A Polyclonal specifically detects KIF21A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KIF21A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CFEOM1, CFEOM3B, FEOM1, FEOM3A, fibrosis of the extraocular muscles, congenital, 1, FLJ20052, KIAA1708DKFZp779C159, KIF2, kinesin family member 21A, Kinesin-like protein KIF2, kinesin-like protein KIF21A, Renal carcinoma antigen NY-REN-62
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1070-1270 of human KIF21A (NP_001166935.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55605
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.