Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kif2a Antibody, Novus Biologicals™
SDP

Catalog No. NB500180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500180 0.1 mL
NB500180SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500180 Supplier Novus Biologicals Supplier No. NB500180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 14 publications

Kif2a Polyclonal specifically detects Kif2a in Bovine, Drosophila melanogaster, Human, Mammalia, Mouse, Ovine, Pig, Rat samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen Kif2a
Applications Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:10000, Immunoprecipitation 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Accession No. O00139
Gene Alias HK2, KIF2kinesin-2, kinesin heavy chain member 2, kinesin heavy chain member 2A, Kinesin, heavy chain, 2, Kinesin-2, kinesin-like protein KIF2A, KNS2
Gene Symbols KIF2A
Host Species Rabbit
Immunogen A recombinant segment of the N-terminal domain of human Kif2a. [UniProt# O00139]
Molecular Weight of Antigen 110 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3796
Target Species Bovine, Drosophila melanogaster, Human, Mammalia, Mouse, Ovine, Pig, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.