Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kif2a Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096207 100 μL
NB096208 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096207 Supplier Novus Biologicals Supplier No. NBP304946100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kif2a Polyclonal specifically detects Kif2a in Rat samples. It is validated for Western Blot.

Specifications

Antigen Kif2a
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias HK2, KIF2kinesin-2, kinesin heavy chain member 2, kinesin heavy chain member 2A, Kinesin, heavy chain, 2, Kinesin-2, kinesin-like protein KIF2A, KNS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-220 of human KIF2A (NP_004511.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3796
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.