Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF7 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291481 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291481 Supplier Novus Biologicals Supplier No. NBP146838PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF7 Polyclonal antibody specifically detects KIF7 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen KIF7
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias EQYK340, kinesin family member 7, kinesin-like protein KIF7, MGC120653, MGC138476, MGC138478, UNQ340
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1225 and 1275 of human kinesin family member 7 using the numbering given in entry NP_940927.2 (GeneID 374654).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 374654
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.