Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIFC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101012 Supplier Invitrogen™ Supplier No. PA5101012
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIFC1.

Minus end-directed microtubule-dependent motor required for bipolar spindle formation (PubMed:15843429). May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIFC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kifc1
Gene Accession No. Q5XI63, Q9BW19, Q9QWT9
Gene Alias 100042970; EMBL:AAH83827.1, ECO:0000312; Gm4137; HSET; Kifc1; kifc1 {ECO:0000312; Kifc4; Kifc5a; Kifc5b; kinesin family c-terminal 5A; kinesin family member C1; kinesin motor protein KIFC4; kinesin-like 2; kinesin-like 2a; kinesin-like protein 2; Kinesin-like protein KIFC1; kinesin-related protein 1; Kinesin-related protein HSET; kinesin-related protein KIFC5A; kinesin-related protein KRP1; KNSL2; Knsl2a; Krp1; RGD:1359118}; Tctex7; Tctex-7; Tctex7a; Tctex-7A
Gene Symbols Kifc1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIFC1(Accession Q9BW19), corresponding to amino acid residues S456-P506.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100502766, 294286, 3833
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.