Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIFC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | KIFC3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KIFC3 Polyclonal specifically detects KIFC3 in Human samples. It is validated for Western Blot.Specifications
| KIFC3 | |
| Polyclonal | |
| Rabbit | |
| Q9BVG8 | |
| 3801 | |
| Synthetic peptides corresponding to KIFC3(kinesin family member C3) The peptide sequence was selected from the C terminal of KIFC3 (NP_005541). Peptide sequence EHLEWEPACQTPQPSARAHSAPSSGTSSRPGSIRRKLQPSGKSRPLPV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp686D23201, FLJ34694, kinesin family member C3, kinesin-like protein KIFC3 | |
| KIFC3 | |
| IgG | |
| 93 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title