Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kilon/NEGR1 Rabbit anti-Mouse, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB081938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081938 Supplier Novus Biologicals Supplier No. NBP298322V

Rabbit Polyclonal Antibody

Kilon/NEGR1 Polyclonal specifically detects Kilon/NEGR1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Kilon/NEGR1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias a kindred of IgLON, IgLON family member 4, IGLON4DMML2433, KILONMGC46680, neuronal growth regulator 1, neurotractin, Ntra
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Kilon/NEGR1 (NP_001034183.1; Met1-Gly318).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257194
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.