Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KleenGuard™ G80 Nitrile Chemical Resistant Gloves
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Hand specific latex-free gloves protect hands against chemical or mechanical hazards
- Diamond embossed fingertips offer enhanced wet and dry grip
- Flock-lined for easy donning and extra comfort in extended use
- Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
- Modified Nitrile formulation for more durability and enhanced chemical resistance.
- Now available in 5 bulk-packed poly bags versus individually wrapped pairs
- Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents

Specifications
Specifications
Certifications/Compliance | FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 |
Certifications/Compliance (Europe) | CE 0120 |
Chlorinated | No |
Color | Green |
Finger Thickness (English) | 15 mil |
Finger Thickness (Metric) | 0.38 mm |
Latex Free | Yes |
Material | Nitrile |
Palm Thickness (English) | 15.7 mil |
Palm Thickness (Metric) | 0.4 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction