Learn More
Ansell KleenGuard™ Nemesis VL™ Safety Glasses
Shop All Ansell Products
Click to view available options
Lens Coating:
Anti-fog
Lens Tint:
Clear
Indoor/Outdoor
Smoke Mirror
Description
Includes
Neck cord
- No-brow design increases upward and peripheral vision
- Slip-stop temples designed to prevent slipping and sliding when hot
- Compact profile accommodates smaller faces
- 99.9% UV protection
- Base curve 8
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (29111-00 meets ANSI Z87.1+2010 and 29111-01 meets ANSI Z87.1+2015) |
| Lens Tint | Clear |
| Frame Color | Gun Metal |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.