Learn More
Ansell KleenGuard™ Nemesis VL™ Safety Glasses
Catalog No. 19706010 Shop All Ansell Products
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor
Smoke Mirror
Lens Coating:
Anti-fog
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Neck cord
Frameless ultra-lightweight design
  • No-brow design increases upward and peripheral vision
  • Slip-stop temples designed to prevent slipping and sliding when hot
  • Compact profile accommodates smaller faces
  • 99.9% UV protection
  • Base curve 8
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1+ standards for high impact (25704-00 meets ANSI Z87.1+2010 and 25704-01 meets ANSI Z87.1+2015)
Lens Tint Smoke Mirror
Frame Color Gun Metal
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.