Learn More
Ansell KleenGuard™ V60 Nemesis™ Safety Eyewear
GREENER_CHOICE
Catalog No. 19005500 Shop All Ansell Products
Click to view available options
Frame Color:
Black
Metallic Blue
Frame Type:
Black frame with black tips
Metallic blue frame with black tips
Lens Tint:
Amber
Clear
Indoor/Outdoor
Smoke
This item is not returnable. View return policy
Catalog No. 19-005-500
Supplier: Ansell 38503
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard Nemesis Safety Eyewear with Rx Safety Inserts are a convenient way to get prescription eyewear into your workplace. They feature the Nemesis Glasses’ sporty good looks and durable, lightweight construction paired with prescription lenses.
  • Fill prescription at a local vision center and fit the inserts into the safety glasses
  • Tend to be more lightweight and comfortable than options like side shields and over-the-glasses safety glasses
  • Polycarbonate lenses help protect the prescription insert from damage and provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Wraparound frame has a base curve of 6.5 that extends the range of peripheral protection, as well as adjustable ratchet-action temples for with five positions for a customized fit and lens angle
  • Corrective lenses are hard coated for scratch resistance and longer life
  • Each pair comes with a neck cord for securing
  • Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
  • Stylish, comfortable and functional for construction, manufacturing, and a host of other industries
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1+ standards for high impact (38503-00 meets ANSI Z87.1+2010 and 38503-01 meets ANSI Z87.1+2015)
Lens Tint Clear
Product Type Safety Eyewear
Frame Color Black
Lens Material Polycarbonate
Frame Type Black frame with black tips
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.