Learn More
KleenGuard™ Nemesis™ Rx Reader Safety Glasses
GREENER_CHOICE
Catalog No. 19170016 Shop All Ansell Products
Click to view available options
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
This item is not returnable. View return policy
Catalog No. 19-170-016
Supplier: KleenGuard™ 28630
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Nemesis™ Rx Reader Safety Glasses are designed for long-term comfort, thanks to flexible and lightweight design, soft touch temples and a nose piece with integrated ridges to channel away sweat
  • Equipped to suit those who need vision assistance
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Wraparound frame has a base curve of 8 that extends the range of peripheral protection, as well as soft touch temples for comfort
  • Corrective lenses are hard coated for scratch resistance and longer life
  • Each pair comes with a neck cord for securing
  • Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
  • Comfortable and functional for construction, manufacturing, and a host of other industries
  • Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose for long-term comfort
Certifications
Meets ANSI Z87.1-2010 Impact standards
CE EN 166: 2001 (Clear lens only); CSA Z94.3-07 (Clear lens only).
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1; CE EN 166: 2001;CSA Z94.3-07
Lens Tint Clear
Product Type Reader Safety Glasses
For Use With (Application) Industrial manufacturing, outdoor environments, impact, splash, grinding/chipping, shaving/sanding
Frame Color Black
Magnification Power +3.00
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.