Kimberly-Clark™ Professional Jackson Saftey™ G40 Blue Foam Nitrile-Coated Gloves

Provide 25% better productivity than leather

Manufacturer: Kimberly-Clark™ Professional 40225

 View more versions of this product

Catalog No. 19-130-3976

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Washable
    Exterior Finish Foam
    Glove Cuff Style Knit
    For Use With (Application) auto parts assembly, materials handling, general maintenance and warehousing
    Length (English) 8.66 in.
    Length (Metric) 21.99cm
    Material Foam Nitrile
    Liner Material Nylon, Spandex™
    Size Small
    Coated Area Palm
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit