Learn More
Ansell KleenGuard™ V80 MonoGoggle™ XTR OTG Safety Goggles
Shop All Ansell Products
Click to view available options
:
MonoGoggle XTR Goggle
Description
- Stylish goggle protects against splashes, while providing increased visibility due to its cylindrical lens
- Soft, pliable frame conforms to the face for comfort and is easy to clean
- Indirect ventilation
- Can be worn over most prescription eyeglasses
- Clear polycarbonate lens with antifog coating
- Blue frame
Specifications
Specifications
Lens Coating | Anti-fog |
Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (18624-02 meets ANSI Z87.1+2010 and 18624-03 meets ANSI Z87.1+2015) with a D3 rating for droplet and splash protection |
Lens Tint | Clear |
Product Type | Safety Goggles; XTR Goggle |
Frame Color | Blue |
Ventilation Details | Indirect |
Material | Polycarbonate |