Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ V80 MRXV™ Safety Goggles

KleenGuard V80 MXRV Safety Goggles give you and your workers reliable splash protection in an unventilated design. These splash goggles have a simple, sleek design and a curved lens for an enhanced vision field, while being roomy enough to fit over most prescription eye glasses.

Specifications

Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Material Polycarbonate
Frame Material PVC
View More Specs

Products 4
Catalog Number Mfr. No. Description Frame Color Price Quantity & Availability  
Catalog Number Mfr. No. Description Frame Color Price Quantity & Availability  
17-986-83D
View Documents
Ansell
16677
VisiClear™ Anti-fog Blue N/A N/A N/A
17-986-83B
View Documents
Ansell
16678
Standard lens Gray N/A N/A N/A
17-986-83E
View Documents
Ansell
16679
VisiClear™ Anti-fog Gray N/A N/A N/A
17-986-83L
View Documents
Ansell
16676
Standard lens Blue N/A N/A N/A
Description

Description

  • Clear, optical-grade polycarbonate lens absorbs 99.9% of UV radiation up to 380nm
  • Oversized PVC frame for a snug, comfortable fit
  • Dual ventilation ports for unobstructed peripheral vision
  • Chemical-resistant neoprene headband attaches to ports on the goggle frame for easy adjustment

Compliance

Meet ANSI™ Z87.1-2010 standards.

Specifications

Specifications

ANSI Z87.1-2010
Clear
Polycarbonate
PVC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.