Learn More
Ansell KleenGuard™ V80 MRXV™ Safety Goggles
Catalog No. 1798683D Shop All Ansell Products
Click to view available options
Description:
Standard lens
VisiClear™ Anti-fog
Frame Color:
Blue
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy
KleenGuard V80 MXRV Safety Goggles give you and your workers reliable splash protection in an unventilated design. These splash goggles have a simple, sleek design and a curved lens for an enhanced vision field, while being roomy enough to fit over most prescription eye glasses.
  • Clear, optical-grade polycarbonate lens absorbs 99.9% of UV radiation up to 380nm
  • Oversized PVC frame for a snug, comfortable fit
  • Dual ventilation ports for unobstructed peripheral vision
  • Chemical-resistant neoprene headband attaches to ports on the goggle frame for easy adjustment
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2010
Description VisiClear™ Anti-fog
Lens Tint Clear
Product Type Safety goggles
Frame Color Blue
Material Polycarbonate
Frame Material PVC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.