Learn More
Ansell KleenGuard™ V80 MonoGoggle™ Safety Goggles
Catalog No. 19706025 Shop All Ansell Products
Click to view available options
Product Type:
Comfort foam frame
Respirator Fit
Smoke frame
Standard frame
Standard frame; Indirect vent
Lens Coating:
Anti-fog
Anti-fog, UV-treated
This item is not returnable. View return policy
Catalog No. 19-706-025
Supplier: Ansell 16670
This item is not returnable. View return policy
Protect your team and yourself from splashes with the KleenGuard V80 Monogoggle. The safety splash goggles give you eye protection from liquids with a two-piece frame which includes a separate soft, cushioning nose piece for enhanced comfort and fit.
  • Standard or fog-resistant VisiClear™ polycarbonate lens provides 99.9% UV protection up to 380nm
  • Patented ventilation system minimizes condensation while internal baffles protect eyes from dust and chemical splash
  • Soft, pliable nosepiece molds to facial contours
  • Headband attaches at the back of the frame for full peripheral vision
  • Standard MonoGoggle with VisiClear with standard PVC frame or with foam-padded frame for additional impact resistance and comfort
  • Respirator-Fit MonoGoggle designed to be worn with a half-mask cartridge or disposable respirator, without inhibiting their protective fit
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2010
Description Fog-resistant VisiClear; Indirect vent
Lens Tint Clear
Product Type Smoke frame
For Use With (Application) Safety
No. per Pack 200/Pk.
Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.