Learn More
Ansell KleenGuard™ V80 SG34 Safety Goggles
Catalog No. 1798683R Shop All Ansell Products
Click to view available options
Description:
Anti-fog Lens
Standard lens
This item is not returnable. View return policy
This item is not returnable. View return policy
When you need comfortable splash protection goggles at an economical price point, look no further than the KleenGuard V80 SG34 Safety Goggles.
  • Polycarbonate lens
  • Button vents prevent indirect venting
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Certifications/Compliance ANSI Z87.1-2010
Description Anti-fog Lens
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Size Universal
Ventilation Details Button vents
Lens Material Polycarbonate
Frame Type Soft, pliable frame for enhanced fit and comfort
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.