Learn More
Ansell KleenGuard™ Visio™ Safety Glasses
Shop All Ansell Products
Click to view available options
Frame Color:
Black
Silver
Lens Tint:
Clear
Clear, Indoor/Outdoor
Description
- One-piece polycarbonate front provides 99.9% UV protection
- Sleek, lightweight, and comfortable
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Uncoated |
| Certifications/Compliance | Meets ANSI Z87.1-2010 standards. |
| Lens Tint | Clear, Indoor/Outdoor |
| Product Type | Safety Glasses |
| Frame Color | Black |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.