Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimtech™ A4 Cleanroom Non-Sterile Sleeve Protectors
GREENER_CHOICE

Catalog No. 19169104 Shop All Ansell Products
Click to view available options
Length (English):
18 in.
21 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Cleanroom sleeve protectors are made with advanced barrier fabric to provide enhanced protection against liquid exposure in cleanroom environments.

  • Made with advanced barrier fabric to provide enhanced protection against liquid exposure
  • Reinforced seams and thumb loops for full wrist coverage
  • Folded inside out for aseptic donning, including a blue indicator line on the interior
  • Double bagged in bulk packaging for convenience and waste reduction
  • Recyclable through the RightCycle Program
  • 200 Kimtech™ A4 Cleanroom Sleeve Protectors/Case, 4 Bags of 50 Sleeves
TRUSTED_SUSTAINABILITY

Specifications

Size Standard
Material Polypropylene
Color White
Sterility Non-sterile
Seam Type Reinforced
Cuff Style Elastic
Model 49815
Includes Thumb Loop
Length (English) 18 in.
Length (Metric) 46 cm
Packaging Double Bagged, Bulk Cleanroom Packaging
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.