Learn More
Kimtech™ A5 Sterile Cleanroom Boots, Grasp Ties and Vinyl Edge
BIOPHARMAGREENER_CHOICE
Catalog No. 19169904 Shop All Ansell Products
Click to view available options
Size:
Small/Medium
Universal
X-Large/2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Sterile cleanroom boots with a wrap-around vinyl foot feature innovative CLEAN-DON™ technology for aseptic donning. Kimtech™ A5 Sterile Cleanroom Boots with Grasp Ties and Vinyl Edge suitable for EU GMP ISO 5 Grade A sterile cleanrooms.
  • Sterile cleanroom boots with grasp ties and vinyl edge
  • Average bacterial filtration efficiency (BFE) of 96%
  • Made of 100% polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-Don ties and 2-inch edge vinyl, for an enhance moisture barrier
  • Made without latex, silicone or the BHT preservative and are designed for single-use
  • 100 Pairs/200 Each
  • CE certified (CE Simple)
  • Each individually bagged pair of boot covers has a sterilization indicator as a visual that it's undergone gamma sterilization
  • A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large/2X-Large
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic with Ties
Sole Type Non-skid
Packaging Quantity 100 Pairs/Cs.
Packaging Type Triple Bagged, Vacuum Packed
Resistance None
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.