Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A10 Light-Duty Coveralls

Catalog No. 19812882 Shop All Ansell Products
Click to view available options
Description:
With Attached Hood and Boots
Without Hood and Boots
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Light-duty coveralls are economically priced for protection in nonhazardous environments.

  • Lightweight, breathable fabric keeps workers cool and comfortable
  • Zipper front
  • Elastic wrists
  • Available with or without attached hood and boots
  • White

Compliance

Meets ANSI/ISEA 101-1996 standards.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Light-duty Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Limited-use
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ANSI/ISEA 101-1996
Description With Attached Hood and Boots
Model KleenGuard™ A10
For Use With (Application) Food Processing, General Maintenance, Manufacturing
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.