Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Lab Coats
GREENER_CHOICE

Catalog No. 19457328 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Closure Type:
Hook and Loop
Snaps
No. of Pockets:
2
None
Cuff Style:
Elastic
Open
This item is not returnable. View return policy

Catalog No. 19-457-328

Supplier: Ansell 35625

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.

  • Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
  • Knee-length, serged seams
  • Long sleeves, notched-lapel collar
  • Air and moisture vapor can pass through to aid wearer comfort
  • Meets NFPA 99 criteria for antistatic materials
  • Two styles to choose from:
    • Open wrists with pockets, snap closure
    • Elastic wrists without pockets, hook and loop closure

Compliance

NFPA 99 compliant antistatic material

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 5X-Large
Gender Unisex
Material SMS
Color White
Closure Type Hook and Loop
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Collar Type Notched Lapel
Model 35625
Seam Type Serged
Certifications/Compliance NFPA 99
For Use With (Application) Laboratories, Remediation, Decontamination, Pesticides
Construction 1.6 oz. Fabric
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.