Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Shirt
Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Designed to help protect against dirt, grime and dust
- Five layers of stacked barrier protection (SMMMS) against hazardous particles
- Hip-length shirt
- Comes in a variety of sizes
- Serged seams
- Five snap closures
- Open wrists
- Mandarin collar
- Popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments
- Made with breathable Microforce barrier fabric
Specifications
Specifications
Size | Large |
Material | Fabric |
Color | White |
Closure Type | Snaps |
Certifications/Compliance | Passes NFPA 99 criteria for antistatic materials |