Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles with Attached Hood and Boots
Shop All Ansell Products

Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
Description
- SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids
- Suitable for many applications including pharmaceutical research, food processing and more
- Protection from non hazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5 cm) flap and elastic back
- Elastic back, wrists and ankles and attached hood and boots

Specifications
Specifications
Product Type | General Purpose Coveralls |
Size | 4X-Large |
Gender | Unisex |
Material | SMS Fabric |
Color | White |
Disposable | Limited-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Attached, Open Face |
Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction