Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A35 Liquid and Particle Protection Coveralls
GREENER_CHOICE

Catalog No. 19006214 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Elastic Wrists and Ankles
Elastic Wrists and Ankles with Attached Hood and Boots
Elastic Wrists, Ankles, and Hood
Open Wrists and Ankles, No Elastic
This item is not returnable. View return policy
This item is not returnable. View return policy

Provide a comfortable fit while providing liquid and particulate protection

  • Breathable
  • Microporous film laminate
  • Lightweight outer fabric that protects the microporous film laminate
  • Added durability and comfort liquid barrier
  • Particulate barrier
  • Available with elastic wrists and ankles
  • With or without hood
  • Zipper front
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust, Liquid
Certifications/Compliance ANSI/ISEA 101-2014
Model KleenGuard™ A35
For Use With (Application) General Industrial, Manufacturing, Laboratory Work, Painting, Maintenance
Style Elastic Wrists and Ankles
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.