Learn More
Ansell KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls
Catalog No. 19057403B Shop All Ansell Products
Click to view available options
Closure Type:
Front Zipper
Zipper with Storm Flap
Description:
Elastic wrist and ankle, seamless zipper Front with storm flap, elastic across the back, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film
Elastic wrist and ankle, seamless zipper with storm flap, elastic across the back, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film
Hooded and booted, seamless zipper front with storm flap, elastic wrist, elastic across the back; 3-ply Fabric construction middle layer of Microporous film
Hooded, seamless zipper front with storm flap, elastic wrist and ankle, elastic across the back; 3-ply Fabric construction middle layer of Microporous film
Seamless zipper front, open wrist and ankle, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
  • Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
  • Feature 12% more chest room and 6% longer body length
  • Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
  • Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
  • Denim blue
Compliance
Meet ASTM F1670/1671 specifications for blood, bodily fluids and bloodborne pathogens.
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Fabric
Color Denim Blue
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Resistance Bloodborne Pathogens, Chemicals
Back Style Elastic
Certifications/Compliance ASTM F1670, 1671
Description Elastic wrist and ankle, seamless zipper Front with storm flap, elastic across the back, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film
For Use With (Application) Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.