Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Lab Coats

Catalog No. 19057440
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Size (English) Chest:
27 in.
28.25 in.
29.75 in.
30 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Lab coats offer bloodborne pathogen and chemical splash protection for applications including forensics and animal research

  • Patented three-layer fabric construction
  • Middle layer is a breathable microporous film that offers resistance against many liquids and dry particulates while also allowing heat and sweat vapor to escape
  • Outer layers are a cloth-like yet tough and abrasion-resistant spunbond polypropylene
  • Snap-front, with chest and hip pocket

Compliance

Fabric passes ASTM F1670/F1671 testing for penetration of blood, bodily fluids, and bloodborne pathogens.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene, Microporous Film
Color Blue
Closure Type Snaps
Disposable Single-use
No. of Pockets 2
Seam Type Serged
Size (English) Chest 27 in.
Certifications/Compliance ASTM F1670, F1671
For Use With (Application) Forensics, Animal Research
Construction 3-ply SMS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.