Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
SDP
Catalog No. p-74781 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
TRUSTED_SUSTAINABILITY
Specifications
Liner Material PTFE, Rubber
Closure Size 8-425
Product Type Screw Cap
Material Phenolic
Color Black
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.