Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin Antibody (4A5.14), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB385357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385357 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385357 Supplier Novus Biologicals Supplier No. NBP308647AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kindlin Monoclonal antibody specifically detects Kindlin in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Kindlin
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4A5.14
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423
Host Species Mouse
Immunogen Recombinant protein containing Kindlin (residues 216-677) (Uniprot: Q9BQL6)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55612
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.