Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000944 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000944 Supplier Novus Biologicals Supplier No. NB100523SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kindlin Polyclonal specifically detects Kindlin in Human samples. It is validated for Western Blot.

Specifications

Antigen Kindlin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9BQL6
Gene Alias C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423
Gene Symbols FERMT1
Host Species Rabbit
Immunogen Reacts with residues 643-660 (VHEYIGGYIFLSTRSKDQ) of the 77 kDa human URP1 protein.
Molecular Weight of Antigen 77 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 55612
Test Specificity URP-1
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.