Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5A Antibody, Novus Biologicals™
SDP

Catalog No. NB1205628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205628 Supplier Novus Biologicals Supplier No. NB1205628

Rabbit Polyclonal Antibody

Kinesin 5A Polyclonal specifically detects Kinesin 5A in Human, Mouse, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kinesin 5A
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml, Flow Cytometry 3 - 5 ug 10^6 cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10
Gene Symbols KIF5A
Host Species Rabbit
Immunogen Synthetic Peptide: C(1007) G Y E A E D Q A K L F P L H Q E T A A S(1027)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3798
Test Specificity This is specific for kinesin 5A and does not detect other kinesin isotypes.
Target Species Human, Mouse, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.