Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5A Antibody, Novus Biologicals™
SDP

Catalog No. NBP232231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232231 Supplier Novus Biologicals Supplier No. NBP232231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kinesin 5A Polyclonal specifically detects Kinesin 5A in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Kinesin 5A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004975.2
Gene Alias D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10
Gene Symbols KIF5A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 982 to 1032 of human Kinesin Family Member 5A using the numbering given in entry NP_004975.2 (GeneID 3798).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3798
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.