Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5B Antibody (KN-02), Novus Biologicals™
SDP

Catalog No. NB500530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500530 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500530 Supplier Novus Biologicals Supplier No. NB500530
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kinesin 5B Monoclonal specifically detects Kinesin 5B in Human, Mouse, Rat, Porcine samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kinesin 5B
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone KN-02
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain
Gene Symbols KIF5B
Host Species Mouse
Immunogen Enriched fraction of porcine brain kinesin.
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3799
Test Specificity The antibody Kinesin 5B recognizes heavy chain of conventional kinesin associated with vesicles and with lower affinity with denaturated molecule. Epitope is located in coiled-coil stalk domain. It stains Western blots of kinesin-enriched preparations. Epitope mapping (by limited proteolysis of partially purified porcine kinesin) followed by immunoblotting has revealed that antibodies KN-01, KN-02 and KN-03 react with different sets of fragments. The antibody KN-02 does not react with kinesin bound to taxol-stabilized microtubules.
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.