Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5B Antibody, Novus Biologicals™
SDP

Catalog No. NBP158177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP158177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP158177 Supplier Novus Biologicals Supplier No. NBP158177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kinesin 5B Polyclonal specifically detects Kinesin 5B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Kinesin 5B
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P33176
Gene Alias Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain
Gene Symbols KIF5B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to KIF5B (kinesin family member 5B) The peptide sequence was selected from the N terminal of KIF5B. Peptide sequence CNIKVMCRFRPLNESEVNRGDKYIAKFQGEDTVVIASKPYAFDRVFQSST.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3799
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Mouse: 92%; Chicken: 85%; Zebrafish: 76%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.