Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5C Antibody, Novus Biologicals™
SDP

Catalog No. NB1205630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205630 Supplier Novus Biologicals Supplier No. NB1205630

Rabbit Polyclonal Antibody

Kinesin 5C Polyclonal specifically detects Kinesin 5C in Human, Bovine (Negative), Mouse (Negative) samples. It is validated for Western Blot.

Specifications

Antigen Kinesin 5C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias heavy chain, neuron-specific, KIF 5C, kinesin family member 5C, Kinesin heavy chain neuron-specific 2, MGC111478, NKHC, NKHC2NKHC-2
Gene Symbols KIF5C
Host Species Rabbit
Immunogen Synthetic Peptide: C A(938) V H A I R G G G G S S S N S T H Y Q K(957)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3800
Test Specificity This is specific for kinesin 5C and does not detect other kinesin isotypes.
Target Species Human, Bovine (Negative), Mouse (Negative)
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.