Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kininogen Antibody (MM0437-11L36), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP211757G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211757G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211757G Supplier Novus Biologicals Supplier No. NBP211757G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kininogen Monoclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kininogen
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0437-11L36
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-2-thiol proteinase inhibitor, BDK, bradykinin, Fitzgerald factor, High molecular weight kininogen, kininogen, kininogen 1, kininogen-1, KNGHMWK, Williams-Fitzgerald-Flaujeac factor
Gene Symbols KNG1
Host Species Mouse
Immunogen Human recombinant Kininogen
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3827
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.