Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL1/CD158a Antibody (MM0438-11G25), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP211758Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211758Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211758Y Supplier Novus Biologicals Supplier No. NBP211758AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR/CD158
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0438-11G25
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Gene Accession No. ACI49719.1
Gene Alias CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor
Gene Symbols KIR2DL1
Host Species Mouse
Immunogen Human recombinant KIR2DL1/CD158a
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3802
Test Specificity This antibody detects KIR2DL2, KIR2DL3, KIR2DS2, and KIR2DS4.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.