Learn More
Description
Specifications
Specifications
| Antigen | KIR2DL1/KIR2DS5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 124 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD158a |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL1/KIR2DS5 (NP_055033.2; Met1-His245). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
