Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL1/KIR2DS5 Antibody (124), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB070712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070712 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070712 Supplier Novus Biologicals Supplier No. NBP290181AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DL1/KIR2DS5
Applications Flow Cytometry
Classification Monoclonal
Clone 124
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD158a
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL1/KIR2DS5 (NP_055033.2; Met1-His245).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3802
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.