Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL4/CD158d Antibody (181703), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB175491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175491 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175491 Supplier Novus Biologicals Supplier No. FAB2238Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR2DL4/CD158d Monoclonal antibody specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry

Specifications

Antigen KIR2DL4/CD158d
Applications Flow Cytometry
Classification Monoclonal
Clone 181703
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL4/CD158d, Accession # Q99706
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3805
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.