Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL4/CD158d Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | KIR2DL4/CD158d |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15460520
![]() |
Novus Biologicals
NBP15460520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154605
![]() |
Novus Biologicals
NBP154605 |
100 μL |
Each for $487.50
|
|
|||||
Description
KIR2DL4/CD158d Polyclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Western Blot.Specifications
| KIR2DL4/CD158d | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
| KIR2DL4 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8NHK1 | |
| 3805 | |
| IgG | |
| 30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title